Learn More
Medchemexpress LLC Cinchonan-9-ol, 10,11-dihydro-6'-methoxy-, (9S)- | 1435-55-8 | 99.97% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB099710G
Encompass_Preferred
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and demonstrates antiarrhythmic efficacy.
  • Strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Demonstrates antiarrhythmic efficacy
  • Has anti-clonogenic and anti-tumorigenic activities on pancreatic cancer cells in vitro
  • For research use only
Catalog No. 50-004-07498