Learn More
Apexbio Technology LLC Cinchonidine 485-71-2 500mg
SDP
Supplier:  Apexbio Technology LLC B3664500
Encompass_Preferred
Cinchonidine (CAS 485-71-2) is a small-molecule inhibitor targeting the serotonin transporter (SERT) It is designed to inhibit SERT function thereby modulating serotonin uptake activity Cinchonidine exerts its biological activity primarily through SERT inhibition In in vitro studies Cinchonidine demonstrates inhibitory activity with IC50 values of 330 M for dSERT and 4 2 36 196 and 15 M for hSERT and its mutants I172M S438T and Y95F respectively Based on these pharmacological properties Cinchonidine holds research potential in serotonin transporter regulation and malaria research
Catalog No. 50-000-90738