Learn More
Medchemexpress LLC Cinchonidine (α-quinidine) | 485-71-2 | 99.6% | 294.39 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN01731G
Encompass_Preferred
Cinchonidine (α-Quinidine) is a cinchona alkaloid found in *Cinchona officinalis* and *Gongronema latifolium*. It is used as a building block in asymmetric synthesis in organic chemistry. It is a weak inhibitor of the serotonin transporter (SERT) and exhibits antimalarial activities.
  • Found in *Cinchona officinalis* and *Gongronema latifolium*.
  • Used as a building block in asymmetric synthesis in organic chemistry.
  • Weak inhibitor of serotonin transporter (SERT).
  • Exhibits antimalarial activities.
  • Physical appearance: Solid, White to off-white.
  • Storage conditions: Room temperature for shipping; 4°C, protected from light, stored under nitrogen for storage.
Catalog No. 50-004-27404