Learn More
Medchemexpress LLC Cinchonidine (α-Quinidine) | 485-71-2 | 99.6% | 294.39 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN0173
Encompass_Preferred
Cinchonidine is a cinchona alkaloid found in plants such as *Cinchona officinalis*. It serves as a building block in asymmetric synthesis and exhibits antimalarial activities.
  • Acts as a weak inhibitor of the serotonin transporter (SERT)
  • Used as a building block in asymmetric synthesis
  • Exhibits antimalarial activities
  • Available in solid, white to off-white appearance
Catalog No. 50-004-29515