Learn More
Medchemexpress LLC Cinnamic acid | 621-82-9 | 148.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN0610A10MM/1ML
Encompass_Preferred
Cinnamic acid is an endogenous metabolite with potential use in cancer intervention, exhibiting effectiveness against glioblastoma, melanoma, prostate, and lung carcinoma cells. It is intended for research use only and not for patient use.
  • Endogenous metabolite
  • Potential use in cancer intervention
  • Effective against glioblastoma, melanoma, prostate, and lung carcinoma cells
  • For research use only
  • Causes morphological changes consistent with melanocyte differentiation
  • Reduces invasiveness in melanoma cells
Catalog No. 50-003-88528