Learn More
Medchemexpress LLC Cinnamic acid-13C3 (3-Phenylacrylic acid-13C3) | 2734410-01-4 | 151.14 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0610AS21MG
Encompass_Preferred
Cinnamic acid-13C3 (3-Phenylacrylic acid-13C3) is a 13C labeled Cinnamic acid, with potential use in cancer intervention. It has shown IC50s of 1-4.5 mM in glioblastoma, melanoma, prostate, and lung carcinoma cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules as tracers for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-88657