Learn More
Medchemexpress LLC Cinnamic acid-13C3 (3-phenylacrylic acid-13C3) | 2734410-01-4 | 151.14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0610AS25MG
Encompass_Preferred
Cinnamic acid-13C3 (3-Phenylacrylic acid-13C3) is the 13C labeled Cinnamic acid. Cinnamic acid has potential use in cancer intervention, with IC50s of 1-4.5 mM in glioblastoma, melanoma, prostate and lung carcinoma cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88627