Learn More
Medchemexpress LLC D-phenylalanine | 673-06-3 | MFCD00004270 | 99.9% | 165.19 | C9H11NO2 | 1g
SDP
Supplier:  Medchemexpress LLC HYY00791G
Encompass_Preferred
D-Phenylalanine is an atypical D-amino acid and an inhibitor of bacterial biofilm formation D-Phenylalanine mainly replaces D-alanine (D-Ala) by incorporating into the fourth and fifth positions of bacterial peptidoglycan (PG) changing the cell wall structure enhancing bacterial acid resistance and affecting biofilm formation D-Phenylalanine may promote the secretion of peptide tyrosine tyrosine (PYY) in mammals by activating the intestinal GPR109B receptor D-Phenylalanine can inhibit the maturation of microbial biofilms and promote the release of specific hormones It can be used for antibacterial preservation improving the yield of probiotics in the food industry and studying appetite regulation and blood sugar control in metabolic diseases such as diabetes[1][2][3][4]
Catalog No. 50-002-69109