Learn More
Selleck Chemical LLC Cinnarizine 100mg 298-57-7 Stugeron, Dimitronal, Stutgin, Cinarizine
SDP
Supplier:  Selleck Chemical LLC S4727100MG
Encompass_Preferred
Cinnarizine (Stugeron, Dimitronal, Stutgin, Cinarizine) is a medication derivative of piperazine, and characterized as an antihistamine and a calcium channel blocker. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4241