Learn More
Medchemexpress LLC Cinnarizine | 298-57-7 | MFCD00056037 | 99.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1090R
Encompass_Preferred
Cinnarizine is provided as a white solid with a molecular formula of C26H28N2 and a molecular weight of 368.51. It is typically stored at -20°C in a sealed brown glass bottle, protected from light and moisture to maintain its stability.
  • White to off-white solid appearance
  • High purity, typically 99.0% by HPLC
  • Consistent with structure as verified by 1H NMR and MS
  • Suitable for laboratory use as a standard
  • Packaged in a brown glass bottle for optimal storage
Catalog No. 50-004-14482