Learn More
Medchemexpress LLC Cinobufagin | 470-37-1 | 442.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN042110MM/1ML
Encompass_Preferred
Cinobufagin is an anticancer agent derived from the Asiatic toad, Bufo gargarizans. It functions by inducing cell cycle arrest in either the G1 or G2/M phases, which subsequently leads to apoptosis in cancer cells. Studies have shown that Cinobufagin can inhibit tumor growth in mouse models with melanoma and glioblastoma multiforme xenografts.
  • Anticancer agent.
  • Induces cell cycle arrest in cancer cells (G1 or G2/M phase).
  • Leads to apoptosis in cancer cells.
  • Inhibits tumor growth in melanoma and glioblastoma multiforme xenograft mouse models.
  • Purity: 99.69%.
Catalog No. 50-003-89952