Learn More
Medchemexpress LLC Cinobufotalin | 1108-68-5 | 458.54 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN08801MG
Encompass_Preferred
Cinobufotalin is a cardiotonic steroid or bufadienolide extracted from the skin secretions of giant toads. It has been used as a cardiotonic, diuretic, and hemostatic agent, and is also a potential anti-lung cancer agent. It is for research use only.
  • Cardiotonic steroid
  • Bufadienolide
  • Extracted from giant toad skin secretions
  • Functions as a cardiotonic, diuretic, and hemostatic agent
  • Potential anti-lung cancer properties
  • For research use only
  • Provided as 1 mg
Catalog No. 50-004-15839