Learn More
Medchemexpress LLC Cinobufotalin | 1108-68-5 | 458.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN088010MM/1ML
Encompass_Preferred
Cinobufotalin is a cardiotonic steroid or bufadienolide extracted from the skin secretions of giant toads. It has been used as a cardiotonic, diuretic, and hemostatic agent, and is also considered a potential anti-lung cancer agent.
  • Cardiotonic steroids or bufadienolides
  • Extracted from the skin secretions of giant toads
  • Potential anti-lung cancer agent
  • High purity
  • For research use only
Catalog No. 50-004-16447