Learn More
Medchemexpress LLC Cinoxacin | 28657-80-9 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB108550MG
Encompass_Preferred
Cinoxacin (Compound 64716) is a synthetic antimicrobial from the quinolone class of orally active antibacterial agents. It exhibits antibacterial activity against many gram-negative aerobic bacteria by inhibiting bacterial DNA synthesis. It is used for research into urinary tract infections and bacterial prostatitis.
  • Exhibits antibacterial activity against gram-negative aerobic bacteria.
  • Inhibits bacterial DNA synthesis.
  • Suitable for research on urinary tract infections.
  • Applicable for research on bacterial prostatitis.
Catalog No. 50-004-12461