Learn More
Medchemexpress LLC Cinoxacin | 28657-80-9 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB108510MM/1ML
Encompass_Preferred
Cinoxacin is a synthetic antimicrobial agent related to the quinolone class of orally active antibacterials. It exhibits antibacterial activity against many gram-negative aerobic bacteria and inhibits bacterial DNA synthesis. Cinoxacin can be used for the research of urinary tract infections and bacterial prostatitis.
  • Synthetic antimicrobial
  • Belongs to the quinolone class
  • Orally active antibacterial agent
  • Inhibits bacterial DNA synthesis
  • Appearance: solid
  • Color: white to off-white
  • Antibacterial activity against many gram-negative aerobic bacteria
  • Can be used for the research of urinary tract infections and bacterial prostatitis
Catalog No. 50-004-11813