Learn More
Medchemexpress LLC Cinoxacin | 28657-80-9 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1085100MG
Encompass_Preferred
Cinoxacin is a synthetic antimicrobial related to the quinolone class of orally active antibacterial agents. It exhibits antibacterial activity against many gram-negative aerobic bacteria and inhibits bacterial DNA synthesis. It can be used for the research of urinary tract infections and bacterial prostatitis.
  • Synthetic antimicrobial
  • Related to the quinolone class
  • Orally active antibacterial agent
  • Inhibits bacterial DNA synthesis
  • Antibacterial activity against many gram-negative aerobic bacteria
  • Can be used for research on urinary tract infections and bacterial prostatitis
Catalog No. 50-004-14059