Learn More
Medchemexpress LLC Cinpa1 | 102636-74-8 | 99.8% | 395.49 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY11024950MG
Encompass_Preferred
CINPA1 is a potent and specific inhibitor of constitutive androstane receptor (CAR) that does not activate pregnane X receptor (PXR). It reduces CAR-mediated transcription and can be used as a molecular tool for understanding CAR function.
  • Inhibits CAR-mediated transactivation without activating PXR in HepG2 cells.
  • Increases corepressor and reduces coactivator interaction with the CAR ligand-binding domain.
  • Disrupts CAR binding to the promoter regions of target genes in chromatin immunoprecipitation assays.
  • Efficiently inhibits CAR-LBD interaction with the coactivator peptide.
Catalog No. 50-003-43960