Learn More
Medchemexpress LLC Cinromide | 58473-74-8 | 99.83% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB127410MM/1ML
Encompass_Preferred
Cinromide is an anticonvulsant agent that inhibits the epithelial neutral amino acid transporter B0AT1 (SLC6A19) with an IC50 of 0.5 μM. It is designed for research use only.
  • Inhibits 5-HT-induced contractions in rat fundus strips by 46% (at 10-100 μM).
  • Inhibits monoamine oxidase from rat liver and brain (at 100 μM).
  • Demonstrates anticonvulsant effects against electroshock and leptazol-induced convulsions in mice.
  • Shows a dose-related antileptazol activity in rats with an ED50 of 58 ± 11 mg/kg.
  • Elevates the amount of leptazol needed to induce clonic seizures in rats.
  • Depresses neuronal response to maxillary nerve stimuli, increasing latency and decreasing spike numbers.
Catalog No. 50-004-12959