Learn More
Medchemexpress LLC Cintirorgon | 2055536-64-4 | MFCD31544352 | 99.1% | 603.53 g/mol | C27H23F6NO6S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10403710MG
Encompass_Preferred
Cintirorgon is a selective, orally bioavailable agonist of the retinoic acid-related orphan receptor gamma (RORγ) developed for research in immuno-oncology. It modulates gene expression in RORγ-expressing T lymphocytes to enhance effector function, reduce immunosuppression, and has shown antitumor activity in preclinical studies.
  • Selective RORγ agonist with targeted immunomodulatory activity.
  • Orally bioavailable for in vivo pharmacology studies.
  • Enhances T cell effector function and reduces immunosuppression.
  • High supplied purity, supporting reliable experimental results.
  • Characterized molecular profile for reproducible research use.
Catalog No. 50-002-44190