Learn More
Medchemexpress LLC Cipargamin | 1193314-23-6 | >98.0% | C19H14Cl2FN3O | 10MG
SDP
Supplier:  Medchemexpress LLC HY1443010MG
Encompass_Preferred
Cipargamin (NITD609) is a synthetic spiroindolone research compound with potent activity against Plasmodium falciparum. It is intended for in vitro and preclinical studies to evaluate antimalarial efficacy and mechanism, and is provided with a defined chemical identity and analytical characterization.
  • Potent antimalarial activity with reported IC50 ≈ 1 nM against Plasmodium falciparum.
  • Spiroindolone class provides a novel mechanism of action for antimalarial research.
  • Suitable for in vitro and preclinical experimental applications.
  • High analytical purity, typically ≥98%.
  • Well-characterized chemical identity and molecular formula C19H14Cl2FN3O.
Catalog No. 50-002-58534