Learn More
Medchemexpress LLC Cipralisant | 213027-19-1 | 99.3% | 216.32 g/mol | C14H20N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10699310MG
Encompass_Preferred
Cipralisant is a small-molecule histamine H3 receptor ligand used in pharmacological research. It acts as a potent, selective H3 receptor antagonist in vivo and can display agonist activity in certain in vitro assays. It is supplied as a purified research-grade compound for use in receptor binding, signaling, and in vivo pharmacology studies.
  • Potent, selective H3 receptor antagonist in vivo.
  • Agonist activity observed in specific in vitro assays.
  • High purity suitable for research applications.
  • Supplied as a purified solid for laboratory studies.
  • Molecular formula C14H20N2 and molecular weight 216.32 g/mol.
Catalog No. 50-002-39767