Learn More
Medchemexpress LLC Cipralisant 5mg | 213027-19-1 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1069935MG
Encompass_Preferred
Cipralisant (GT-2331) is an orally active low-toxicity potent selective high affinity histamine H3 receptor full antagonist in vivo and an agonist in vitro with a pKi of 9 9 for histamine H3 receptor and a Ki of 0 47 nM for rat histamine H3 receptor Cipralisant has the potential for attention-deficit hyperactivity disorder research[1 [2 [3 [4 Cipralisant is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-39712