Learn More
Medchemexpress LLC Cipralisant (enantiomer) | 223420-11-9 | 100.0% | 216.32 g/mol | C14H20N2 | 1 ML

Supplier: Medchemexpress LLC HY106993B10MM 1ML
Cipralisant (enantiomer) is the enantiomeric form of cipralisant supplied as a 10 mM solution in DMSO for research use. It is intended for in vitro pharmacology and analytical applications related to histamine H3 receptor studies and is not for human or clinical use.
- 10 mM solution in DMSO for ready-to-use experiments.
- 1 mL sealed vial suitable for small-scale assays.
- Molecular weight 216.32 g/mol for accurate calculations.
- High enantiomeric excess (~96.4% ee) to support stereospecific studies.
- Supplier-stated purity 100% for analytical consistency.
- Characterization data available (COA, HNMR, NP-HPLC, LCMS) to support quality control.
- Intended for in vitro H3 receptor pharmacology and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.