Learn More
Enzo Life Sciences Ciprofibrate (25mg). CAS: 52214-84-3
SDP
Supplier:  Enzo Life Sciences ALX270475M025
Encompass_Preferred
Hypolipidemic compound. PPARα (peroxisome proliferator-activated receptor α) agonist. Alternative name: 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8600