Learn More
Cayman Chemical CIprofIbrate-d6 5mg
SDP
Supplier:  Cayman Chemical 339175 MG
Encompass_Preferred
An internal standard for the quantification of ciprofibrate by GC- or LC-MS
Catalog No. 50-294-8075