Learn More
Apexbio Technology LLC Ciprofloxacin (hydrochloride)(Synonyms: Cipro, Ciprofloxacin HCl, Ciprofloxacine hydrochloride, Ciprofloxacin hydrochloride monohydrate), 50g, CAS: 93107-08-5.
SDP
Supplier:  Apexbio Technology LLC C553950000
Encompass_Preferred
Ciprofloxacin (hydrochloride) (CAS 93107-08-5) is a fluoroquinolone antibiotic targeting bacterial DNA gyrase and topoisomerase IV enzymes essential for bacterial DNA replication and supercoiling By inhibiting these enzymes ciprofloxacin prevents bacterial chromosome replication and proliferation It also displays immunomodulatory activity evidenced by reduced serum pro-inflammatory cytokines (IL-6 KC) and decreased apoptosis and autophagy in radiation-induced injury mouse models Ciprofloxacin is FDA-approved for treating inhalational anthrax exposure significantly enhancing survival in rhesus monkeys infected with aerosolized B anthracis
Catalog No. 50-000-61300