Learn More
Apexbio Technology LLC Ciprofloxacin 85721-33-1 5g
SDP
Supplier:  Apexbio Technology LLC A83995000
Encompass_Preferred
Ciprofloxacin (85721-33-1) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) and topoisomerase IV It is designed to inhibit these enzymes thereby disrupting bacterial DNA replication and transcription Ciprofloxacin exerts its biological activity primarily through inhibition of DNA gyrase and topoisomerase IV In in vitro and cell-based studies Ciprofloxacin demonstrates inhibitory activity with IC50 values ranging from approximately 0 1 g/mL to 2 5 g/mL depending on the bacterial strain and assay conditions Based on these pharmacological properties Ciprofloxacin holds research potential in studies of DNA enzymology bacterial gene regulation and antimicrobial resistance mechanisms
Catalog No. 50-000-96065