Learn More
Medchemexpress LLC Ciprofloxacin | 85721-33-1 | 99.8% | 331.34 | 500 MG
SDP
Supplier:  Medchemexpress LLC 500MGHYB0356
Encompass_Preferred
Ciprofloxacin (Bay-09867) is a potent, orally active topoisomerase IV inhibitor. It induces mitochondrial DNA and nuclear DNA damage, leading to mitochondrial dysfunction and ROS production. Ciprofloxacin exhibits anti-proliferative activity and induces apoptosis. It is also a fluoroquinolone antibiotic with potent antibacterial activity.
  • Potent, orally active topoisomerase IV inhibitor
  • Induces mitochondrial and nuclear DNA damage
  • Leads to mitochondrial dysfunction and ROS production
  • Exhibits anti-proliferative activity
  • Induces apoptosis
  • Potent antibacterial activity
Catalog No. 50-003-65726