Learn More
Medchemexpress LLC Ciprofloxacin-d8 hydrochloride | 1216659-54-9 | 98.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0356S1MG
Encompass_Preferred
Ciprofloxacin-d8 hydrochloride is the deuterium labeled Ciprofloxacin. This fluoroquinolone antibiotic exhibits potent antibacterial activity.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06875