Learn More
Medchemexpress LLC Ciprofloxacin-d8 (hydrochloride) | 1216659-54-9 | 98.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0356S5MG
Encompass_Preferred
Ciprofloxacin-d8 (hydrochloride) is a deuterium labeled fluoroquinolone antibiotic, exhibiting potent antibacterial activity. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits potent antibacterial activity
Catalog No. 50-004-05377