Learn More
Bioworld Ciprofloxacin (Cipro) Hydrochloride, Ready-to-Use, Sterile, 25 mL
Supplier:  Bioworld 403100312
Encompass_Preferred
Ciprofloxacin Hydrochloride (Cipro-HCl)For Mycoplasma/Bacterial Elimination from animal cell cultures.1000X, 10mg/mL,effective concentration 10ug/ml in most culture media.ml> false false false ml> Mycoplasma contamination is a major problem in cell culture in biomedical and cell/molecular biology research. There is no easy visual confirmation of mycoplasma contamination which can have serious effects on key characteristics and functions of a cell line, thus compromising all results obtained using such cultures. Cipro HCl is the standard antibiotic for rapid and efficient removal of mycoplasma from contaminated cultures. Bio-World offers 99% pure Cipro in a ready-to-use 1000X solution or in powder form for quick elimination of this problem.Applications: -Treatment of mycoplasma-infected cell lines -Decontamination of visible Pen-Strep resistant bacteria -Prevent contamination from plasmid preparation during transfections.
Catalog No. 50-255-730