Learn More
Abcam Ciproflo x acin hydrochloride, Fluoroquinolone antibiotic, 5G
Supplier:  Abcam AB1419185G
Encompass_Preferred
MW 385.8 Da. Fluoroquinolone antibiotic. Bacteriostatic agent. Interferes with DNA gyrase causing DNA rewinding stopping DNA and protein synthesis. Active in vivo and in vitro..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6401