Learn More
Aviva Systems Biology Corp CIR Antibody - C-terminal region (ARP38523_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38523_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CIR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CIR. Alternate names: CIRApplicationWB. Species reactivity: Human
Catalog No. 50-014-56685