Learn More
Grainger Circuit Breaker 250A 3P 600VAC JG
Supplier:  Grainger 14G981
Encompass_Preferred
The Grainger Approved 1-Pole 15 Amp 120/277 Volt DIN Rail Mount Circuit Breaker is a 1-pole circuit breaker with a continuous current rating of 15A at 120/277V AC. It features a thermal-magnetic trip unit that provides overload and short circuit protection. This circuit breaker has a maximum voltage rating of 480V AC and mounts onto 35mm DIN rails. It has a width of 18mm. The circuit breaker is UL listed and CSA certified. This DIN rail mount breaker is ideal for industrial control panels and machinery requiring circuit protection.
Catalog No. 50-275-0849
Add to Cart
Request a Quote