Learn More
Grainger Circuit Breaker 60A 3P 600VAC HG
Supplier:  Grainger 14G969
Encompass_Preferred
The 14G969 Circuit Breaker is a 60A 3P 600VAC HG circuit breaker manufactured by Schneider Electric. It features a thermal-magnetic trip unit to provide overload and short circuit protection. This 3 pole circuit breaker has a maximum voltage rating of 600VAC and a continuous current rating of 60 amps. It conforms to IEC 60947-2 standards with a 10kA breaking capacity. The circuit breaker has screw type terminals and can be easily installed into load centers or panelboards. It provides reliable circuit protection for commercial and industrial applications.
Catalog No. 50-275-3486