Learn More
Medchemexpress LLC Cirsimarin | 13020-19-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN72725MG
Encompass_Preferred
Cirsimarin is a potent antilipogenic flavonoid isolated from Microtea debilis. It exerts a potent antilipogenic effect and decreases adipose tissue deposition in mice. Its lipolytic activity results from both its antagonist activity on adenosine A1 receptor and its inhibitory effect on phosphodiesterase.
  • Potent antilipogenic flavonoid
  • Exerts a potent antilipogenic effect
  • Phosphodiesterase (PDE) inhibitor
  • Decreases adipose tissue deposition in mice
  • For research use only
Catalog No. 50-004-18555