Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cirsimaritin | 6601-62-3 | 99.2% | 314.29 g/mol | C17H14O6 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN664810MG

Encompass_Preferred

Cirsimaritin is a flavone-class natural product used as a pharmacological research tool. It binds weakly to the benzodiazepine site on GABAA receptors and has reported antidepressant, anxiolytic, and antinociceptive activities.

  • High chemical purity reported at 99.23%.
  • CAS number 6601-62-3 for unambiguous identification.
  • Molecular weight 314.29 g/mol, formula C17H14O6.
  • Soluble in DMSO (22.73 mg/mL) and formulatable in 10% DMSO/90% corn oil for in vivo use.
  • Offered in small research sizes suitable for discovery work, including 10 mg and larger packs.
  • Powder stable at -20°C (up to 3 years); store solutions at -80°C for long-term stability.

Catalog No. 50-002-89774


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.