Learn More
Medchemexpress LLC cis-1-Benzyl-3,5-dimethyl-4-oxopiperidine | 324769-03-1 | 217.31 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW004217
cis-1-Benzyl-3,5-dimethyl-4-oxopiperidine is a drug intermediate for the synthesis of various active compounds. It appears as a colorless to light yellow liquid.
- Product type: Drug intermediate
- Appearance: Liquid
- Color: Colorless to light yellow
- Shipment at room temperature in continental US
- Storage for pure form: -20°C for 3 years, 4°C for 2 years
- Storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.