Learn More
AdipoGen cis-11-Methyl-dodecenoic acid
SDP
Supplier:  AdipoGen CDXM0075M010
Encompass_Preferred
Chemical. CAS 677354-23-3. Formula C13H24O2. MW 212.33. Synthetic. A diffusible signal factor DSF in extracellular microbial and fungal communication systems. In a DSF bioassay, the minimum concentration of cis-Delta2-11-methyl-dodecenoic acid required for induction of a DSF biosensor was about 0.5 µM, which is 200-fold less than that of the conformational isomer trans-Delta2-11-methyl-dodecenoic acid and 20,000-fold lower than that of the corresponding saturated fatty acid 11-methyl-dodecanoic acid.
Catalog No. 50-256-0544