Learn More
AdipoGen cis-2-Decenoic acid
SDP
Supplier:  AdipoGen CDXD0249G001
Encompass_Preferred
Chemical. CAS 15790-91-7. Formula C10H18O2. MW 170.25. Synthetic. cis-2-Decenoic acid is an unsaturated short chain fatty acid that is secreted by P. aeruginosa and induces a dispersion response in biofilms formed by gram-negative and gram-positive bacteria, as well as by the yeast C. albicans.
Catalog No. 50-256-1168