Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC cis-2-Hexen-1-ol | 928-94-9 | 91.3% | 100.16 | 1 G
SDP

Supplier:  Medchemexpress LLC 1GHYW127618

Encompass_Preferred

cis-2-Hexen-1-ol is a biochemical reagent suitable for life science-related research. This organic compound is intended for research use only.

  • Used as a biological material or organic compound
  • Suitable for life science-related research
  • Liquid appearance
  • Colorless to light yellow
  • Formula: C6H12O
  • Recommended storage for pure form: -20°C for 3 years

Catalog No. 50-003-65199


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.