Learn More
Medchemexpress LLC Cis-2-(methoxycarbonyl)cyclopentanecarboxylic acid | 92541-43-0 | MFCD01311173 | 98.0% | C8H12O4 | 250 MG

Supplier: Medchemexpress LLC 250MGHY76450
Cis-2-(methoxycarbonyl)cyclopentanecarboxylic acid is a laboratory chemical with the molecular formula C8H12O4. This compound is an irritant and may cause skin irritation, serious eye irritation, and respiratory irritation. It is stable under recommended storage conditions.
- For laboratory chemical use
- Stable under recommended storage conditions
- Purity of 98.0% (NMR)
- Appears as a yellow to brown liquid
- Store at -20°C for 3 years or 4°C for 2 years in pure form
- Store at -80°C for 6 months or -20°C for 1 month in solvent
- IUPAC name: cis-(1R,2S)-2-methoxycarbonylcyclopentane-1-carboxylic acid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.