Learn More
Medchemexpress LLC Cis-2-penten-1-ol | 1576-95-0 | 86.13 g/mol | C5H10O | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0303475G
Encompass_Preferred
cis-2-Penten-1-ol (CAS 1576-95-0) is an unsaturated primary alcohol used as an organic reagent and building block in research and synthesis. Supplied in small laboratory pack sizes for analytical and preparative work, it is suitable for use in flavor and fragrance studies, method development, and organic transformations. Formula: C5H10O; molecular weight: 86.13 g/mol.
  • Unsaturated primary alcohol suitable for organic synthesis and derivatization.
  • Available in small laboratory pack sizes for research-scale use.
  • Useful as a reference standard in analytical and chromatographic methods.
  • Characteristic fruity/green odor useful in flavor and fragrance studies.
  • Simple molecular structure facilitates downstream chemical transformations.
Catalog No. 50-003-00238