Learn More
Medchemexpress LLC Cis-3,4',5-trimethoxy-3'-hydroxystilbene | 586410-08-4 | 99.6% | 286.33 g/mol | C17H18O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11845250MG
Encompass_Preferred
cis-3,4',5-Trimethoxy-3'-hydroxystilbene is a cis-stilbene research compound (C17H18O4, MW 286.33 g/mol) supplied as an oil. It is used in cellular studies of cytochrome c release and inhibition of tubulin polymerization; analytical data and certificates are available.
Catalog No. 50-002-27440