Learn More
Selleck Chemical LLC cis-4-Hydroxy-D-proline S3367-100mg
SDP
Supplier:  Selleck Chemical LLC S3367100MG
Encompass_Preferred
cis-4-Hydroxy-D-proline (H-D-cis-Hyp-OH) is an endogenous metabolite that can be used to study the specificity and kinetics of D-alanine dehydrogenase
Catalog No. 50-311-0139