Learn More
Medchemexpress LLC cis-4-Hydroxy-L-proline | 618-27-9 | >=98.0% | 131.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY40136S5MG
Encompass_Preferred
cis-4-Hydroxy-L-proline is an orally active proline analog and a collagen production inhibitor. It inhibits cell growth by preventing the deposition of triple helical collagen on the cell layer. This product has been shown to inhibit N-nitrosomethylurea-induced breast tumor growth and can improve lung compliance while reducing prostate weight. It is also suitable for use in the study of hypoxic pulmonary hypertension.
  • Orally active proline analog
  • Collagen production inhibitor
  • Inhibits cell growth by preventing triple helical collagen deposition
  • Inhibits N-nitrosomethylurea-induced breast tumor growth
  • Improves lung compliance
  • Reduces prostate weight
  • Can be used in the study of hypoxic pulmonary hypertension
Catalog No. 50-004-03787