Learn More
Medchemexpress LLC cis-4-Hydroxy-L-proline | 618-27-9 | MFCD00064319 | 98.0% | 131.13 | 1 G
SDP
Supplier:  Medchemexpress LLC HY401361G
Encompass_Preferred
cis-4-Hydroxy-L-proline is an orally active proline analog that acts as a collagen production inhibitor. It prevents the deposition of triple helical collagen on the cell layer, thereby inhibiting cell growth. This compound has been shown to inhibit N-nitrosomethylurea-induced breast tumor growth, improve lung compliance, and reduce prostate weight. It is useful in studies related to hypoxic pulmonary hypertension.
  • Orally active proline analog
  • Inhibits collagen production and cell growth
  • Inhibits N-nitrosomethylurea-induced breast tumor growth
  • Improves lung compliance and reduces prostate weight
  • Used in studies of hypoxic pulmonary hypertension
  • Demonstrates in vitro inhibition of collagenous and noncollagenous protein accumulation
  • Reduces collagen synthesis in chick tendon fibroblasts in vitro
  • Partially prevents collagen accumulation in lungs in vivo
Catalog No. 50-004-04653