Learn More
Medchemexpress LLC cis-5-Octen-1-ol 10mM 1mL
SDP
Supplier:  Medchemexpress LLC HYW10130110MM/1ML
Encompass_Preferred
cis-5-Octen-1-ol is an organic compound belonging to alcohols It has a strong earthy smell and is commonly found in a variety of foods such as fruits vegetables and spices cis-5-Octen-1-ol has various applications in the flavor and fragrance industry especially as a flavoring and aroma enhancer in products such as perfume cologne and baked goods In addition it has potential utility in inhibiting inflammation-related diseases and cancer
Catalog No. 50-002-05651