Learn More
Cayman Chemical cIs-6-HexadecenoIc AcId 5mg
SDP
Supplier:  Cayman Chemical 90018455 MG
Encompass_Preferred
A monounsaturated fatty acid; one of the primary fatty acids in human skin; levels are increased in isolated sebum from the face and back of patients with acne; levels are decreased in the non-lesional skin and isolated sebum of atopic dermatitis patients; active against S. aureus in vitro at 5 UG/ml at pH 5.5
Catalog No. 50-289-4875