Learn More
Cayman Chemical cIs-8-OctadecenoIc AcId 5mg
SDP
Supplier:  Cayman Chemical 274475 MG
Encompass_Preferred
A monounsaturated fatty acid; an isomer of oleic acid, trans-vaccenic acid, trans-petroselinic acid, and cis-petroselinic acid; has been found in partially hydrogenated vegetable oil and milk fat
Catalog No. 50-293-8278